Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321995.1 | NP_001308924.1 | 5 | 623 | ||
NM_031902.4 | NP_114108.1 | 5 | 802 | |||
NR_136153.1 | 5 | 802 | ||||
GenBank mRNA | AB049940.1 | - | 5 | 683 | ||
AK058160.1 | - | 5 | 576 | |||
AK295812.1 | - | 5 | 588 | |||
BC000219.2 | - | 1 | 117 | |||
BC014172.2 | - | 5 | 775 | 4561399 |