Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271667.1 | NP_001258596.1 | 6 | 608 | ||
NM_001271668.1 | NP_001258597.1 | 5 | 512 | |||
NM_001271669.1 | NP_001258598.1 | 4 | 458 | |||
NM_032122.4 | NP_115498.2 | 6 | 563 | |||
NM_183040.2 | NP_898861.1 | 6 | 563 | |||
NR_036448.1 | 7 | 702 | ||||
XM_005249447.4 | XP_005249504.1 | 7 | 564 | |||
XM_011514936.2 | XP_011513238.1 | 6 | 554 | |||
GenBank mRNA | AF061734.1 | - | 2 | 50 | ||
AF394226.1 | - | 6 | 480 | |||
AK054593.1 | - | 6 | 532 | |||
AK290718.1 | - | 6 | 480 | |||
AK310590.1 | - | 7 | 564 | |||
AL136637.2 | - | 6 | 480 | |||
AY265460.1 | - | 6 | 487 | |||
BC011912.2 | - | 6 | 478 | 4139934 | ||
BE793298.1 | - | 420 | ||||
BQ056746.1 | - | 5 | 451 | |||
BX394617.2 | - | 7 | 702 | |||
KU178801.1 | - | 6 | 374 | |||
KU178802.1 | - | 4 | 269 |