Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242832.1 | NP_001229761.1 | 11 | 1273 | ||
NM_032389.4 | NP_115765.2 | 12 | 1357 | |||
XM_005253166.1 | XP_005253223.1 | 13 | 1852 | |||
XM_005253167.1 | XP_005253224.1 | 13 | 1819 | |||
XM_005253168.2 | XP_005253225.1 | 13 | 1777 | |||
XM_006718346.1 | XP_006718409.1 | 12 | 1810 | |||
XM_011520405.1 | XP_011518707.1 | 12 | 1693 | |||
XM_017018413.1 | XP_016873902.1 | 12 | 1777 | |||
XM_017018414.1 | XP_016873903.1 | 12 | 1173 | |||
XM_017018415.1 | XP_016873904.1 | 12 | 1125 | |||
GenBank mRNA | AB073358.1 | - | 11 | 967 | ||
AK027482.1 | - | 12 | 1093 | |||
AK098324.1 | - | 7 | 703 | |||
AK129511.1 | - | 2 | 50 | |||
AK301785.1 | - | 11 | 977 | |||
AK304541.1 | - | 12 | 932 | |||
AK315992.1 | - | 6 | 624 | |||
AL834337.1 | - | 12 | 1076 | |||
BC030148.2 | - | 12 | 1100 | 4558453 | ||
BC036823.1 | - | 12 | 1102 |