Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003417.4 | NP_003408.1 | 2 | 545 | ||
XM_011527522.2 | XP_011525824.1 | 2 | 545 | |||
GenBank mRNA | AB007872.1 | - | 2 | 494 | ||
AK292504.1 | - | 2 | 545 | |||
BC115410.2 | - | 2 | 187 | |||
BC115411.1 | - | 3 | 227 | 40078437 | ||
BM739537.1 | - | 2 | 544 |
Gene Symbol: ZNF805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023563.3 | NP_001018857.2 | 2 | 335 |