Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143774.1 | NP_001137246.1 | 4 | 586 | ||
NM_032439.3 | NP_115815.2 | 4 | 806 | |||
XM_011540275.2 | XP_011538577.1 | 5 | 868 | |||
XM_011540276.2 | XP_011538578.1 | 4 | 1699 | |||
XM_017016782.1 | XP_016872271.1 | 5 | 687 | |||
XM_017016783.1 | XP_016872272.1 | 5 | 1790 | |||
GenBank mRNA | AB058699.1 | - | 2 | 147 | ||
AK054956.1 | - | 4 | 610 | |||
AL365474.1 | - | 4 | 586 | |||
AL834339.1 | - | 4 | 807 | |||
AY358162.1 | - | 3 | 436 | |||
BC011268.2 | - | 4 | 645 | 4156912 | ||
CR749429.1 | - | 5 | 835 |