Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201484.1 | NP_001188413.1 | 7, 8 | 1618 | ||
NM_032634.3 | NP_116023.2 | 6, 7 | 1520 | |||
NM_152850.3 | NP_690577.2 | 6, 7 | 1166 | |||
XM_005251619.3 | XP_005251676.1 | 6, 7 | 1145 | |||
XM_017015222.1 | XP_016870711.1 | 7, 8 | 1637 | |||
XM_017015223.1 | XP_016870712.1 | 6, 7 | 1547 | |||
XM_017015224.1 | XP_016870713.1 | 6, 7 | 1811 | |||
GenBank mRNA | AL833956.1 | - | 6, 7 | 1556 | ||
AY358472.1 | - | 5, 6 | 1126 | |||
BC001030.2 | - | 3, 4 | 496 | |||
BC013987.2 | - | 4, 5 | 582 | |||
BC029271.1 | - | 5, 6 | 791 | |||
BC036916.1 | - | 5, 6 | 1131 | 5203414 | ||
BC065282.1 | - | 7, 8 | 1289 | 6172460 |