Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADTRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143948.1 | NP_001137420.1 | 4 | 698 | ||
NM_032744.3 | NP_116133.1 | 3 | 644 | |||
XM_005249454.3 | XP_005249511.1 | 4 | 669 | |||
XM_011514956.1 | XP_011513258.1 | 4 | 669 | |||
XM_011514958.2 | XP_011513260.1 | 4 | 669 | |||
XM_011514959.1 | XP_011513261.1 | 4 | 669 | |||
XM_017011377.1 | XP_016866866.1 | 3 | 262 | |||
GenBank mRNA | AJ420520.1 | - | 3 | 413 | ||
AK300919.1 | - | 4 | 454 | |||
AK313114.1 | - | 3 | 400 | |||
BC007011.1 | - | 3 | 430 | 3686576 |