Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032826.4 | NP_116215.1 | 8 | 980 | ||
XM_011516645.2 | XP_011514947.1 | 8 | 748 | |||
GenBank mRNA | AB052892.1 | - | 8 | 956 | ||
AJ315497.1 | - | 8 | 812 | |||
AJ315498.1 | - | 8 | 802 | |||
AJ971941.1 | - | 8 | 656 | |||
AK027603.1 | - | 8 | 820 | |||
AK027726.1 | - | 7 | 538 | |||
AK075368.1 | - | 8 | 785 | |||
AK222874.1 | - | 8 | 829 | |||
AK307621.1 | - | 7 | 758 | |||
BC008413.1 | - | 8 | 801 | 4333393 |