Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145160.1 | NP_001138632.1 | 9 | 2213 | ||
NM_003290.2 | NP_003281.1 | 8 | 2228 | |||
XM_011528231.2 | XP_011526533.1 | 9 | 2126 | |||
XM_011528232.1 | XP_011526534.1 | 8 | 3446 | |||
XM_017027188.1 | XP_016882677.1 | 9 | 2244 | |||
XM_017027189.1 | XP_016882678.1 | 8 | 4079 | |||
GenBank mRNA | FJ224305.1 | - | 8 | 2037 | ||
X05276.1 | - | 8 | 2037 |