Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DIRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032839.2 | NP_116228.1 | 9 | 1702 | ||
GenBank mRNA | AK027690.1 | - | 9 | 1696 | ||
AK075158.1 | - | 10 | 1714 | |||
BC022204.1 | - | 9 | 1960 | |||
BC033738.1 | - | 8 | 1578 | |||
BC039821.1 | - | 9 | 1673 | 6062886 |