Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCAY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033064.4 | NP_149053.1 | 13 | 2085 | ||
GenBank mRNA | AB058775.1 | - | 13 | 2029 | ||
AK027889.1 | - | 13 | 2019 | |||
AK092309.1 | - | 13 | 1884 | |||
AK125457.1 | - | 13 | 1606 | |||
BC026217.1 | - | 13 | 2027 | 4153341 |