Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNKS1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033396.2 | NP_203754.2 | 5 | 1585 | ||
XM_006718725.3 | XP_006718788.1 | 5 | 1601 | |||
XM_011545325.2 | XP_011543627.1 | 5 | 1598 | |||
GenBank mRNA | AB051528.2 | - | 5 | 1565 | ||
AF441771.1 | - | 4 | 1727 | |||
AK124903.1 | - | 5 | 1612 | |||
AK127871.1 | - | 3 | 1008 | |||
BC150333.1 | - | 5 | 1576 | 8860508 |