Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277126.1 | NP_001264055.1 | 10 | 3498 | ||
NM_001277129.1 | NP_001264058.1 | 9 | 3324 | |||
NM_144687.3 | NP_653288.1 | 10 | 3495 | |||
XM_011527479.1 | XP_011525781.1 | 9 | 3327 | |||
XM_011527480.1 | XP_011525782.1 | 9 | 3327 | |||
XM_011527482.1 | XP_011525784.1 | 8 | 3156 | |||
XM_017027460.1 | XP_016882949.1 | 10 | 3495 | |||
XM_017027461.1 | XP_016882950.1 | 9 | 3327 | |||
XM_017027462.1 | XP_016882951.1 | 9 | 3324 | |||
XM_017027463.1 | XP_016882952.1 | 11 | 3271 | |||
XM_017027464.1 | XP_016882953.1 | 10 | 3069 | |||
XM_017027465.1 | XP_016882954.1 | 11 | 3279 | |||
XM_017027466.1 | XP_016882955.1 | 11 | 3276 | |||
XM_017027467.1 | XP_016882956.1 | 10 | 3071 | |||
GenBank mRNA | AF231021.1 | - | 7 | 2252 | ||
AK095460.1 | - | 9 | 3239 | |||
AY095146.1 | - | 10 | 3442 | |||
AY116204.1 | - | 10 | 3489 | |||
AY116205.1 | - | 9 | 3321 | |||
AY116206.1 | - | 8 | 3153 | |||
AY116207.1 | - | 7 | 2979 | |||
AY154467.1 | - | 10 | 3442 | |||
BC028069.1 | - | 10 | 3385 | 5212737 |