Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACBD5 SIRNA Assays ›
Gene Symbol:
ACBD5
Gene Name:
acyl-CoA binding domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.10: 27182624 - 27248674 on Build GRCh38
UniGene ID:
Hs.530597
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131804
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACBD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042473.3
NP_001035938.1
8
944
NM_001271512.3
NP_001258441.1
7
1258
NM_001301251.1
NP_001288180.1
7
823
NM_001301252.1
NP_001288181.1
6
1137
NM_001301253.1
NP_001288182.1
7
813
NM_001301254.1
NP_001288183.1
5
933
NM_145698.4
NP_663736.2
8
1069
XM_006717528.3
XP_006717591.3
9
7502
XM_006717530.3
XP_006717593.1
9
1101
XM_006717531.2
XP_006717594.1
9
1111
XM_006717535.2
XP_006717598.1
9
968
XM_017016884.1
XP_016872373.1
9
7502
XM_017016885.1
XP_016872374.1
8
7469
XM_017016886.1
XP_016872375.1
8
7469
XM_017016887.1
XP_016872376.1
9
1032
XM_017016888.1
XP_016872377.1
9
1097
XM_017016889.1
XP_016872378.1
8
999
XM_017016890.1
XP_016872379.1
8
7298
XM_017016891.1
XP_016872380.1
8
1064
XM_017016892.1
XP_016872381.1
8
985
XM_017016893.1
XP_016872382.1
7
7265
XM_017016894.1
XP_016872383.1
7
7265
XM_017016895.1
XP_016872384.1
9
968
XM_017016896.1
XP_016872385.1
8
936
XM_017016897.1
XP_016872386.1
8
893
XM_017016898.1
XP_016872387.1
7
860
XM_017016899.1
XP_016872388.1
7
860
XM_017016900.1
XP_016872389.1
8
886
XM_017016901.1
XP_016872390.1
8
866
XM_017016902.1
XP_016872391.1
8
885
XM_017016903.1
XP_016872392.1
8
870
XM_017016904.1
XP_016872393.1
7
865
XM_017016905.1
XP_016872394.1
7
851
XM_017016906.1
XP_016872395.1
7
732
XM_017016907.1
XP_016872396.1
7
865
XM_017016908.1
XP_016872397.1
7
854
GenBank mRNA
AB082527.1
-
7
772
AF505653.1
-
7
810
AK057469.1
-
8
936
AK294510.1
-
5
851
AK295031.1
-
7
895
AK304842.1
-
6
788
AL133064.1
-
2
152
BC030555.1
-
8
973
5218685
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
macroautophagy
pexophagy
aggrephagy
fatty-acyl-CoA binding
molecular_function
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0