Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACBD5 SIRNA Assays ›
Gene Symbol:
ACBD5
Gene Name:
acyl-CoA binding domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.10: 27182624 - 27248674 on Build GRCh38
UniGene ID:
Hs.530597
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131806
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACBD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042473.3
NP_001035938.1
13
1836
NM_001271512.3
NP_001258441.1
12
2150
NM_001301251.1
NP_001288180.1
12
1715
NM_001301252.1
NP_001288181.1
11
2029
NM_001301253.1
NP_001288182.1
12
1705
NM_001301254.1
NP_001288183.1
10
1825
NM_145698.4
NP_663736.2
13
1961
XM_006717528.3
XP_006717591.3
14
8394
XM_006717530.3
XP_006717593.1
14
1993
XM_006717531.2
XP_006717594.1
14
2003
XM_006717535.2
XP_006717598.1
14
1860
XM_017016886.1
XP_016872375.1
13
8361
XM_017016892.1
XP_016872381.1
13
1877
XM_017016894.1
XP_016872383.1
12
8157
XM_017016897.1
XP_016872386.1
13
1785
XM_017016899.1
XP_016872388.1
12
1752
XM_017016902.1
XP_016872391.1
13
1777
XM_017016903.1
XP_016872392.1
13
1762
XM_017016906.1
XP_016872395.1
12
1624
XM_017016907.1
XP_016872396.1
12
1757
XM_017016908.1
XP_016872397.1
12
1746
GenBank mRNA
AB082527.1
-
12
1664
AF505653.1
-
12
1702
AK057469.1
-
13
1828
AK294510.1
-
10
1743
AK304842.1
-
11
1680
AL133064.1
-
7
1044
BC030555.1
-
13
1865
5218685
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
macroautophagy
pexophagy
aggrephagy
fatty-acyl-CoA binding
molecular_function
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0