Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005781.1 | NP_001005781.1 | 5 | 405 | ||
NM_001005782.1 | NP_001005782.1 | 4 | 330 | |||
NM_003352.4 | NP_003343.1 | 5 | 405 | |||
GenBank mRNA | AB062294.1 | - | 5 | 381 | ||
AK311840.1 | - | 5 | 370 | |||
BC005899.1 | - | 5 | 372 | |||
BC006462.1 | - | 5 | 388 | 3546125 | ||
BC053528.1 | - | 5 | 363 | 3452929 | ||
BC066306.1 | - | 5 | 381 | 4818792 | ||
BG569821.1 | - | 4 | 298 | |||
BT006632.1 | - | 1 | 257 | |||
BU532734.1 | - | 354 | ||||
CR542147.1 | - | 1 | 257 | |||
CR542156.1 | - | 1 | 257 | |||
U38784.1 | - | 6 | 323 | |||
U61397.1 | - | 5 | 402 | |||
U67122.1 | - | 5 | 337 | |||
U72722.1 | - | 1 | 257 | |||
U83117.1 | - | 5 | 392 | |||
X99586.1 | - | 298 |
Gene Symbol: SUMO1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002190.1 | 1 | 393 |