Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCUBE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173050.3 | NP_766638.2 | 5, 6 | 735 | ||
XM_005261752.1 | XP_005261809.1 | 5, 6 | 696 | |||
XM_011530389.2 | XP_011528691.1 | 5, 6 | 690 | |||
XM_011530390.2 | XP_011528692.1 | 4, 5 | 575 | |||
GenBank mRNA | AF525689.1 | - | 6, 7 | 628 | ||
AK127719.1 | - | 5, 6 | 696 | |||
BC047916.1 | - | 5, 6 | 674 | |||
BC127263.1 | - | 5, 6 | 632 |