Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003358.2 | NP_003349.1 | 5, 6 | 1016 | ||
XM_005252186.4 | XP_005252243.1 | 6, 7 | 1039 | |||
GenBank mRNA | AK314847.1 | - | 5, 6 | 621 | ||
BC038711.1 | - | 5, 6 | 1017 | 5295561 | ||
D50840.1 | - | 5, 6 | 856 |