Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WDR20 SIRNA Assays ›
Gene Symbol:
WDR20
Gene Name:
WD repeat domain 20
Gene Aliases:
DMR
Chromosome Location:
Chr.14: 102139518 - 102224686 on Build GRCh38
UniGene ID:
Hs.36859
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131869
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WDR20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242414.1
NP_001229343.1
3
849
NM_001242416.1
NP_001229345.1
2
836
NM_001242417.1
NP_001229346.1
4
1112
NM_001242418.1
NP_001229347.1
3
1055
NM_144574.3
NP_653175.2
3
1019
NM_181291.2
NP_851808.1
3
1019
NM_181308.2
NP_851825.1
2
836
NR_038354.1
4
812
XM_006720308.1
XP_006720371.1
2
828
XM_006720309.1
XP_006720372.1
3
802
XM_006720310.1
XP_006720373.1
3
763
XM_006720311.1
XP_006720374.1
2
592
XM_006720313.2
XP_006720376.1
4
771
XM_006720318.2
XP_006720381.1
3
710
XM_011537335.2
XP_011535637.1
4
1131
XM_011537336.2
XP_011535638.1
4
1131
XM_011537337.2
XP_011535639.1
4
1131
XM_011537338.1
XP_011535640.1
4
1078
XM_011537339.2
XP_011535641.1
4
1131
XM_011537340.2
XP_011535642.1
4
1131
XM_011537341.2
XP_011535643.1
3
1037
XM_011537343.1
XP_011535645.1
5
1106
XM_011537344.2
XP_011535646.1
4
1024
XM_011537345.1
XP_011535647.1
3
802
XM_011537346.1
XP_011535648.1
3
828
XM_011537347.1
XP_011535649.1
4
826
XM_011537353.2
XP_011535655.1
4
882
XM_011537358.2
XP_011535660.1
3
787
XM_017021766.1
XP_016877255.1
4
1078
XM_017021767.1
XP_016877256.1
3
1037
XM_017021768.1
XP_016877257.1
3
1052
XM_017021770.1
XP_016877259.1
3
1052
XM_017021771.1
XP_016877260.1
4
1087
XM_017021772.1
XP_016877261.1
4
840
XM_017021773.1
XP_016877262.1
5
933
XM_017021774.1
XP_016877263.1
4
882
XM_017021775.1
XP_016877264.1
4
888
XM_017021776.1
XP_016877265.1
3
802
XM_017021777.1
XP_016877266.1
3
770
XM_017021778.1
XP_016877267.1
4
882
XM_017021779.1
XP_016877268.1
2
592
XM_017021780.1
XP_016877269.1
4
1003
XM_017021781.1
XP_016877270.1
3
710
XM_017021782.1
XP_016877271.1
3
523
XM_017021783.1
XP_016877272.1
4
819
XM_017021785.1
XP_016877274.1
4
1078
GenBank mRNA
AB209148.1
-
1
5108
AF327354.1
-
3
944
AK056330.1
-
3
761
AK090580.1
-
4
771
AK090978.1
-
3
790
AK225852.1
-
2
795
AK297727.1
-
4
1050
AL133558.1
-
1
256
AL832242.1
-
3
788
AL832500.1
-
3
994
BC028387.2
-
3
978
4823662
BX248274.1
-
2
738
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0