Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015423.2 | NP_056238.2 | 6 | 1759 | ||
GenBank mRNA | AB072896.1 | - | 1 | 698 | ||
AF151838.1 | - | 6 | 1778 | |||
AF302110.1 | - | 6 | 1779 | |||
AL050073.1 | - | 5 | 1320 | |||
BX537665.1 | - | 2 | 4039 |