Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243746.1 | NP_001230675.1 | 1 | 493 | ||
NM_017565.3 | NP_060035.2 | 1 | 493 | |||
XM_011524918.2 | XP_011523220.1 | 1 | 333 | |||
XM_017024781.1 | XP_016880270.1 | 1 | 333 | |||
GenBank mRNA | AB545606.1 | - | 1 | 493 | ||
AK056789.1 | - | 1 | 333 | |||
AK298071.1 | - | 1 | 135 | |||
AY358197.1 | - | 1 | 493 | |||
BC136686.1 | - | 1 | 382 | 9020676 | ||
BC136689.1 | - | 1 | 382 | 9020679 |