Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243746.1 | NP_001230675.1 | 10 | 1758 | ||
NM_017565.3 | NP_060035.2 | 9 | 1532 | |||
NR_027751.1 | 9 | 959 | ||||
XM_006721959.2 | XP_006722022.1 | 9 | 904 | |||
GenBank mRNA | AB545606.1 | - | 9 | 1532 | ||
AF447872.1 | - | 3 | 1345 | |||
AK056789.1 | - | 10 | 1372 | |||
AK298071.1 | - | 10 | 1400 | |||
AL133105.1 | - | 7 | 641 | |||
AY358197.1 | - | 10 | 1532 | |||
BC036222.1 | - | 9 | 923 | |||
BC136686.1 | - | 9 | 1421 | 9020676 | ||
BC136689.1 | - | 10 | 1421 | 9020679 |