Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017634.3 | NP_060104.2 | 2 | 349 | ||
GenBank mRNA | AK298518.1 | - | 2 | 153 | ||
BC021216.2 | - | 2 | 152 | 4302317 | ||
BC068518.1 | - | 2 | 162 | 5265435 | ||
BG249272.1 | - | 287 | ||||
BU149129.1 | - | 2 | 267 | |||
DA115324.1 | - | 2 | 350 |