Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEKHB2 SIRNA Assays ›
Gene Symbol:
PLEKHB2
Gene Name:
pleckstrin homology domain containing B2
Gene Aliases:
EVT2
Chromosome Location:
Chr.2: 131104847 - 131149852 on Build GRCh38
UniGene ID:
Hs.469944
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132203
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEKHB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100623.1
NP_001094093.1
3
706
NM_001267062.1
NP_001253991.1
3
706
NM_001267063.1
NP_001253992.1
3
706
NM_001267064.1
NP_001253993.1
3
706
NM_001267065.1
NP_001253994.1
3
706
NM_001267066.1
NP_001253995.1
3
706
NM_001267067.1
NP_001253996.1
3
706
NM_001267068.1
NP_001253997.1
3
706
NM_001309448.1
NP_001296377.1
2
661
NM_001309450.1
NP_001296379.1
3
706
NM_001309451.1
NP_001296380.1
2
661
NM_001309452.1
NP_001296381.1
4
804
NM_017958.2
NP_060428.2
3
706
XM_011511385.2
XP_011509687.1
3
245
XM_011511386.2
XP_011509688.1
3
248
XM_017004408.1
XP_016859897.1
2
174
XM_017004409.1
XP_016859898.1
2
174
XM_017004410.1
XP_016859899.1
2
174
XM_017004411.1
XP_016859900.1
4
1310
XM_017004412.1
XP_016859901.1
4
1310
XM_017004413.1
XP_016859902.1
3
224
GenBank mRNA
AK000790.1
-
3
217
AK093730.1
-
3
941
AK123710.1
-
3
663
AK126289.1
-
3
706
AK223338.1
-
3
217
AK293880.1
-
3
217
AK296008.1
-
2
170
AK302770.1
-
3
251
BC001428.1
-
3
200
3139876
BC013991.2
-
3
199
3639839
BC050695.1
-
3
201
6162960
BP362174.1
-
2
172
BQ052811.1
-
3
198
BQ649140.1
-
3
215
BU174451.1
-
4
285
CK000843.1
-
3
209
DB294071.1
-
3
219
DC303684.1
-
3
224
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell differentiation
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cnw2s:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0