Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304496.1 | NP_001291425.1 | 6 | 1038 | ||
NM_018073.7 | NP_060543.5 | 7 | 1521 | |||
GenBank mRNA | AF360739.1 | - | 7 | 1474 | ||
AF439153.1 | - | 6 | 1231 | |||
AK001231.1 | - | 4 | 726 | |||
AK022923.1 | - | 4 | 691 | |||
AK056523.1 | - | 7 | 1483 | |||
AK291166.1 | - | 7 | 1433 | |||
AK301120.1 | - | 5 | 753 | |||
AL832730.1 | - | 7 | 1471 | |||
BC075058.2 | - | 6 | 1255 | 30915367 | ||
BC109063.2 | - | 7 | 1440 | 40033795 |