Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132296
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077351.1
NP_001070819.1
8
817
NM_001077352.1
NP_001070820.1
6
715
NM_001308044.1
NP_001294973.1
6
656
NM_018107.4
NP_060577.3
7
769
XM_011536890.2
XP_011535192.1
9
900
XM_011536892.2
XP_011535194.1
8
851
XM_011536893.2
XP_011535195.1
8
852
XM_011536894.2
XP_011535196.1
8
846
XM_011536895.2
XP_011535197.1
7
822
XM_011536896.2
XP_011535198.1
7
798
XM_011536897.2
XP_011535199.1
7
1184
XM_011536900.1
XP_011535202.1
7
760
XM_011536902.1
XP_011535204.1
7
666
XM_011536903.1
XP_011535205.1
5
542
XM_011536904.1
XP_011535206.1
6
599
XM_011536905.1
XP_011535207.1
5
597
XM_011536906.1
XP_011535208.1
4
488
XM_017021398.1
XP_016876887.1
8
854
XM_017021399.1
XP_016876888.1
7
801
XM_017021400.1
XP_016876889.1
7
719
XM_017021401.1
XP_016876890.1
8
814
XM_017021402.1
XP_016876891.1
8
725
XM_017021403.1
XP_016876892.1
7
743
XM_017021404.1
XP_016876893.1
7
693
XM_017021405.1
XP_016876894.1
7
760
XM_017021406.1
XP_016876895.1
8
1232
XM_017021407.1
XP_016876896.1
8
741
XM_017021408.1
XP_016876897.1
6
677
XM_017021409.1
XP_016876898.1
7
1178
XM_017021410.1
XP_016876899.1
6
699
XM_017021411.1
XP_016876900.1
6
1130
XM_017021412.1
XP_016876901.1
8
852
XM_017021413.1
XP_016876902.1
7
628
XM_017021414.1
XP_016876903.1
7
666
XM_017021415.1
XP_016876904.1
6
647
XM_017021416.1
XP_016876905.1
5
542
XM_017021417.1
XP_016876906.1
6
599
XM_017021418.1
XP_016876907.1
5
597
XM_017021419.1
XP_016876908.1
4
488
XM_017021420.1
XP_016876909.1
5
1017
GenBank mRNA
AF087905.1
-
7
720
AF275678.1
-
8
772
AK001344.1
-
6
665
AK125593.1
-
8
779
AK225792.1
-
8
779
AK303668.1
-
6
618
AK308055.1
-
7
732
AK315620.1
-
7
731
AL834198.1
-
8
778
BC002566.2
-
7
747
3138909
BC024208.2
-
7
706
2960467
BC106012.1
-
5
491
BX161440.1
-
8
750
CR457257.1
-
6
570
KU178687.1
-
6
570
KU178688.1
-
7
618
KU178689.1
-
5
516
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
nucleotide binding
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0