Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R3 SIRNA Assays ›
Gene Symbol:
PPP6R3
Gene Name:
protein phosphatase 6 regulatory subunit 3
Gene Aliases:
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location:
Chr.11: 68460712 - 68615334 on Build GRCh38
UniGene ID:
Hs.503022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132427
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164160.1
NP_001157632.1
3
468
NM_001164161.1
NP_001157633.1
3
468
NM_001164162.1
NP_001157634.1
3
468
NM_001164163.1
NP_001157635.1
3
468
NM_001164164.1
NP_001157636.1
4
573
NM_018312.4
NP_060782.2
4
573
XM_006718608.3
XP_006718671.1
4
584
XM_006718612.3
XP_006718675.1
4
584
XM_006718613.2
XP_006718676.1
4
584
XM_006718614.2
XP_006718677.1
4
584
XM_006718618.3
XP_006718681.1
4
584
XM_006718621.2
XP_006718684.1
4
584
XM_006718622.3
XP_006718685.1
4
584
XM_006718623.3
XP_006718686.1
4
567
XM_006718624.2
XP_006718687.1
4
584
XM_006718627.3
XP_006718690.1
4
584
XM_011545135.1
XP_011543437.1
4
584
XM_011545136.2
XP_011543438.1
3
467
XM_011545137.2
XP_011543439.1
4
567
XM_011545138.2
XP_011543440.1
5
785
XM_011545139.1
XP_011543441.1
4
584
XM_011545140.2
XP_011543442.1
4
584
XM_011545141.1
XP_011543443.1
4
584
XM_011545142.2
XP_011543444.1
4
584
XM_011545143.1
XP_011543445.1
4
584
XM_011545145.2
XP_011543447.1
4
584
XM_017017962.1
XP_016873451.1
3
422
XM_017017963.1
XP_016873452.1
4
508
XM_017017964.1
XP_016873453.1
4
584
XM_017017965.1
XP_016873454.1
3
467
XM_017017966.1
XP_016873455.1
4
670
XM_017017967.1
XP_016873456.1
4
584
XM_017017968.1
XP_016873457.1
3
467
XM_017017969.1
XP_016873458.1
4
584
XM_017017970.1
XP_016873459.1
3
467
XM_017017971.1
XP_016873460.1
4
584
XM_017017972.1
XP_016873461.1
3
440
XM_017017973.1
XP_016873462.1
4
567
XM_017017974.1
XP_016873463.1
4
584
XM_017017975.1
XP_016873464.1
4
584
XM_017017976.1
XP_016873465.1
3
467
XM_017017983.1
XP_016873472.1
4
587
XM_017017984.1
XP_016873473.1
3
404
GenBank mRNA
AB046778.2
-
3
416
AF264779.1
-
3
478
AF264780.1
-
3
478
AK315136.1
-
3
437
BC105933.1
-
2
336
40029064
BC105934.1
-
2
336
40029058
BC107599.1
-
2
336
40029065
BP314824.1
-
3
468
CN351527.1
-
4
542
CR749815.1
-
3
431
CR933658.1
-
3
431
CR936718.1
-
3
436
DQ111954.2
-
1
201
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of phosphoprotein phosphatase activity
COPII vesicle coating
protein binding
protein phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0