Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300926.1 | NP_001287855.1 | 7 | 1065 | ||
NM_018320.4 | NP_060790.2 | 9 | 1227 | |||
NR_024147.1 | 8 | 1189 | ||||
NR_024148.1 | 9 | 1327 | ||||
XM_006718629.1 | XP_006718692.1 | 8 | 1140 | |||
XM_006718630.1 | XP_006718693.1 | 10 | 1040 | |||
XM_006718632.1 | XP_006718695.1 | 7 | 985 | |||
XM_011545150.1 | XP_011543452.1 | 10 | 1072 | |||
GenBank mRNA | AK001961.1 | - | 4 | 391 | ||
AK023139.1 | - | 9 | 893 | |||
AK094508.1 | - | 9 | 993 | |||
AL833638.1 | - | 7 | 740 | |||
BC009672.2 | - | 9 | 895 | |||
BC063680.1 | - | 4 | 403 | |||
BC093084.1 | - | 8 | 829 | |||
BC117273.1 | - | 9 | 859 | |||
BG328761.1 | - | 3 | 199 | |||
BX463021.2 | - | 5 | 494 | |||
CR457298.1 | - | 9 | 887 |