Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300926.1 | NP_001287855.1 | 7 | 1116 | ||
NM_018320.4 | NP_060790.2 | 9 | 1278 | |||
NR_024147.1 | 8 | 1240 | ||||
NR_024148.1 | 9 | 1378 | ||||
XM_006718629.1 | XP_006718692.1 | 8 | 1191 | |||
XM_006718630.1 | XP_006718693.1 | 10 | 1091 | |||
XM_006718632.1 | XP_006718695.1 | 7 | 1036 | |||
XM_011545150.1 | XP_011543452.1 | 10 | 1123 | |||
GenBank mRNA | AK001961.1 | - | 4 | 442 | ||
AK023139.1 | - | 9 | 944 | |||
AK094508.1 | - | 9 | 1044 | |||
AL833638.1 | - | 7 | 791 | |||
BC009672.2 | - | 9 | 946 | |||
BC063680.1 | - | 4 | 454 | |||
BC093084.1 | - | 8 | 880 | |||
BC117273.1 | - | 9 | 910 | |||
BG328761.1 | - | 3 | 250 | |||
BX463021.2 | - | 5 | 545 | |||
CR457298.1 | - | 9 | 938 |