Gene Symbol: NIPSNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018376.3 | NP_060846.2 | 6 | 1446 | ||
NR_130759.1 | 5 | 1235 | ||||
XM_011518839.1 | XP_011517141.1 | 5 | 1296 | |||
GenBank mRNA | AK002137.1 | - | 6 | 1051 | ||
AK310397.1 | - | 5 | 791 | |||
BC005202.1 | - | 6 | 979 | 3933738 | ||
BC017914.1 | - | 6 | 1017 | 4292375 |