Gene Symbol: STK32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306082.1 | NP_001293011.1 | 13 | 2162 | ||
NM_018401.2 | NP_060871.1 | 12 | 2052 | |||
XM_005247982.3 | XP_005248039.1 | 7 | 2559 | |||
XM_005247983.4 | XP_005248040.1 | 7 | 1817 | |||
XM_011513497.1 | XP_011511799.1 | 11 | 2156 | |||
XM_011513501.2 | XP_011511803.1 | 8 | 1351 | |||
XM_011513504.1 | XP_011511806.1 | 7 | 1205 | |||
XM_017008377.1 | XP_016863866.1 | 7 | 2604 | |||
GenBank mRNA | AJ250839.1 | - | 12 | 1769 | ||
AK055597.1 | - | 3 | 696 | |||
AK292696.1 | - | 13 | 1828 | |||
AL832436.1 | - | 6 | 1105 | |||
AY358353.1 | - | 12 | 1796 | |||
BC020281.1 | - | 1 | 111 | |||
BC024977.1 | - | 1 | 111 | |||
BC038238.1 | - | 12 | 1746 | 5744969 |