Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018431.4 | NP_060901.2 | 2 | 595 | ||
NM_177959.2 | NP_808874.1 | 2 | 241 | |||
XM_011528904.1 | XP_011527206.1 | 2 | 224 | |||
GenBank mRNA | AF132732.1 | - | 2 | 317 | ||
AF466368.1 | - | 2 | 120 | |||
AK313222.1 | - | 2 | 307 | |||
BC008992.1 | - | 2 | 241 | 4183000 | ||
BI758267.1 | - | 2 | 340 |