Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282529.1 | NP_001269458.1 | 19 | 2374 | ||
NM_001282530.1 | NP_001269459.1 | 7 | 917 | |||
NM_018449.3 | NP_060919.3 | 23 | 2688 | |||
GenBank mRNA | AB040924.1 | - | 11 | 1490 | ||
AK025357.1 | - | 4 | 330 | |||
AK025801.1 | - | 12 | 1578 | |||
AK026088.1 | - | 15 | 1856 | |||
AK057564.1 | - | 4 | 1243 | |||
AK074884.1 | - | 10 | 1047 | |||
AK125935.1 | - | 6 | 2172 | |||
AK128424.1 | - | 1 | 544 | |||
AK294952.1 | - | 23 | 2606 | |||
AK299183.1 | - | 17 | 2096 | |||
AK302754.1 | - | 22 | 2556 | |||
AK303493.1 | - | 19 | 2312 | |||
BC037416.1 | - | 4 | 321 |