Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018688.4 | NP_061158.1 | 9 | 1067 | ||
XM_005273573.3 | XP_005273630.1 | 8 | 973 | |||
XM_011544586.2 | XP_011542888.1 | 10 | 1152 | |||
GenBank mRNA | AF271732.1 | - | 9 | 1030 | ||
AK001289.1 | - | 7 | 929 | |||
AK023980.1 | - | 9 | 1014 | |||
AK124486.1 | - | 3 | 1352 | |||
BC001223.1 | - | 8 | 951 | 3534031 | ||
BC009824.2 | - | 9 | 1001 | 3953604 |