Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303145.1 | NP_001290074.1 | 2 | 904 | ||
NM_018939.3 | NP_061762.2 | 1 | 1109 | |||
GenBank mRNA | AF152499.1 | - | 1 | 873 | ||
AF217752.1 | - | 1 | 873 | |||
AK299661.1 | - | 2 | 827 | |||
AK313482.1 | - | 1 | 1030 | |||
DA386733.1 | - | 1 | 36 |