Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146265.1 | NP_001139737.1 | 3 | 1943 | ||
NM_001146266.1 | NP_001139738.1 | 2 | 1833 | |||
NM_001146267.1 | NP_001139739.1 | 3 | 1770 | |||
NM_018970.6 | NP_061843.3 | 3 | 1940 | |||
XM_005250451.2 | XP_005250508.1 | 2 | 1890 | |||
XM_017012352.1 | XP_016867841.1 | 3 | 2005 | |||
GenBank mRNA | AF250237.2 | - | 3 | 1928 | ||
AK297464.1 | - | 4 | 998 | |||
AK302769.1 | - | 2 | 725 | |||
AL161959.1 | - | 3 | 1773 | |||
BC030577.1 | - | 3 | 1894 | 4823671 |
Set Membership: |
![]() |