Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018983.3 | NP_061856.1 | 2, 3 | 520 | ||
NM_032993.2 | NP_127460.1 | 2, 3 | 261 | |||
GenBank mRNA | AJ276003.1 | - | 2, 3 | 477 | ||
AK310489.1 | - | 2, 3 | 403 | |||
AY780787.1 | - | 1, 2 | 213 | |||
BC003413.1 | - | 2, 3 | 261 | 3451136 | ||
CN428426.1 | - | 2, 3 | 485 |