Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242581.1 | NP_001229510.1 | 12 | 4291 | ||
NM_018993.3 | NP_061866.1 | 11 | 4031 | |||
XM_005260731.2 | XP_005260788.1 | 13 | 4487 | |||
XM_005260733.2 | XP_005260790.1 | 9 | 3839 | |||
XM_006723574.3 | XP_006723637.1 | 14 | 4469 | |||
XM_006723575.3 | XP_006723638.1 | 13 | 4255 | |||
XM_006723577.2 | XP_006723640.1 | 12 | 4104 | |||
XM_011529255.2 | XP_011527557.2 | 12 | 4564 | |||
XM_011529257.1 | XP_011527559.1 | 12 | 4160 | |||
XM_011529258.2 | XP_011527560.1 | 12 | 4172 | |||
XM_017027887.1 | XP_016883376.1 | 15 | 4810 | |||
XM_017027888.1 | XP_016883377.1 | 16 | 4882 | |||
XM_017027889.1 | XP_016883378.1 | 11 | 4411 | |||
XM_017027890.1 | XP_016883379.1 | 13 | 4242 | |||
GenBank mRNA | AL136924.1 | - | 10 | 3910 |