Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288607.1 | NP_001275536.1 | 11 | 2024 | ||
NM_001288608.1 | NP_001275537.1 | 11 | 2024 | |||
NM_019032.5 | NP_061905.2 | 10 | 1955 | |||
NM_025008.4 | NP_079284.2 | 10 | 1955 | |||
XM_011509644.2 | XP_011507946.1 | 10 | 1986 | |||
XM_011509645.2 | XP_011507947.1 | 9 | 1914 | |||
XM_011509648.2 | XP_011507950.1 | 10 | 1940 | |||
XM_011509649.2 | XP_011507951.1 | 10 | 1994 | |||
XM_011509650.2 | XP_011507952.1 | 10 | 1994 | |||
XM_011509651.2 | XP_011507953.1 | 4 | 736 | |||
XM_011509652.1 | XP_011507954.1 | 4 | 442 | |||
XM_017001506.1 | XP_016856995.1 | 11 | 2395 | |||
GenBank mRNA | AF370411.1 | - | 4 | 491 | ||
AK092509.1 | - | 1 | 75 | |||
AY358122.1 | - | 9 | 1769 | |||
BC027478.1 | - | 3 | 394 | |||
BC071852.1 | - | 2 | 251 | |||
BC094811.1 | - | 2 | 251 | |||
BC140800.1 | - | 10 | 1876 | 9021668 | ||
BC144581.1 | - | 10 | 1876 | 9053111 |