Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301088.1 | NP_001288017.1 | 12 | 1678 | ||
NM_019055.5 | NP_061928.4 | 12 | 1884 | |||
XM_006718861.1 | XP_006718924.1 | 12 | 1817 | |||
XM_011542875.1 | XP_011541177.1 | 6 | 646 | |||
GenBank mRNA | AF361473.1 | - | 12 | 1813 | ||
AK000805.1 | - | 4 | 340 | |||
AK027852.1 | - | 12 | 1783 | |||
AK074163.1 | - | 11 | 1414 | |||
AK289769.1 | - | 12 | 1783 | |||
AK297845.1 | - | 6 | 646 | |||
AK302628.1 | - | 12 | 1548 | |||
AY358083.1 | - | 12 | 1735 | |||
BC014995.1 | - | 11 | 1496 | |||
BC025675.1 | - | 2 | 676 | |||
BC039602.1 | - | 12 | 1772 | 5590503 | ||
BC111562.1 | - | 12 | 1778 | 40068748 | ||
BC111748.1 | - | 12 | 1778 | 40068752 |