Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301088.1 | NP_001288017.1 | 18 | 3168 | ||
NM_019055.5 | NP_061928.4 | 18 | 3374 | |||
XM_006718861.1 | XP_006718924.1 | 18 | 3193 | |||
XM_011542875.1 | XP_011541177.1 | 12 | 2136 | |||
GenBank mRNA | AF361473.1 | - | 18 | 3303 | ||
AK025195.1 | - | 6 | 1224 | |||
AK027852.1 | - | 18 | 3272 | |||
AK074163.1 | - | 15 | 4232 | |||
AK297845.1 | - | 12 | 1954 | |||
AY358083.1 | - | 18 | 3225 | |||
BC014995.1 | - | 17 | 2986 | |||
BC025675.1 | - | 5 | 2697 | |||
BC039602.1 | - | 18 | 3262 | 5590503 | ||
BC064643.1 | - | 8 | 1521 | |||
BC111562.1 | - | 18 | 3268 | 40068748 | ||
BC111748.1 | - | 18 | 3268 | 40068752 |