Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279349.1 | NP_001266278.1 | 2 | 541 | ||
NM_001279350.1 | NP_001266279.1 | 2 | 541 | |||
NM_001279351.1 | NP_001266280.1 | 2 | 402 | |||
NM_020385.3 | NP_065118.2 | 2 | 719 | |||
GenBank mRNA | AF273304.1 | - | 2 | 667 | ||
AF295774.1 | - | 2 | 667 | |||
AK025493.1 | - | 2 | 573 | |||
AK295982.1 | - | 2 | 402 | |||
AK304547.1 | - | 2 | 457 | |||
AK314333.1 | - | 2 | 633 | |||
AK315904.1 | - | 3 | 392 | |||
AL136894.1 | - | 2 | 650 | |||
BC009274.2 | - | 2 | 622 | 3958331 | ||
BX333016.2 | - | 2 | 431 |