Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014447.2 | NP_001014447.1 | 7 | 1268 | ||
NM_001014448.2 | NP_001014448.1 | 7 | 1531 | |||
NM_003652.3 | NP_003643.2 | 6 | 1235 | |||
GenBank mRNA | AK056317.1 | - | 7 | 1267 | ||
AK127844.1 | - | 4 | 2604 | |||
BC006393.2 | - | 7 | 1435 | |||
BC018913.2 | - | 7 | 1435 | |||
BC080539.1 | - | 7 | 1202 | 5020194 | ||
U83411.1 | - | 6 | 1100 |