Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242924.1 | NP_001229853.1 | 12 | 1861 | ||
NM_001242926.1 | NP_001229855.1 | 10 | 1669 | |||
NM_001242927.1 | NP_001229856.1 | 10 | 1591 | |||
NM_001242928.1 | NP_001229857.1 | 11 | 1781 | |||
NM_021188.2 | NP_067011.1 | 11 | 1810 | |||
NR_040251.1 | 11 | 1778 | ||||
GenBank mRNA | AK223261.1 | - | 11 | 1566 | ||
AK293350.1 | - | 12 | 1617 | |||
AK298307.1 | - | 12 | 1533 | |||
AK298520.1 | - | 11 | 1586 | |||
AK299138.1 | - | 10 | 1589 | |||
AK303834.1 | - | 10 | 1270 | |||
AK304609.1 | - | 11 | 1324 | |||
AK309957.1 | - | 12 | 1606 | |||
AK315206.1 | - | 11 | 1568 | |||
BC004357.1 | - | 11 | 1525 | |||
BC050683.1 | - | 10 | 1479 | 5752452 | ||
U90919.1 | - | 11 | 1542 |