Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172673.1 | NP_001166144.1 | 12 | 4699 | ||
NM_021938.3 | NP_068757.2 | 12, 13 | 1545 | |||
NR_033342.1 | 12, 13 | 1342 | ||||
XM_006722832.1 | XP_006722895.1 | 13 | 3286 | |||
XM_006722833.1 | XP_006722896.1 | 13 | 3283 | |||
XM_006722836.2 | XP_006722899.1 | 12 | 2927 | |||
XM_006722837.3 | XP_006722900.1 | 13 | 3198 | |||
XM_006722838.3 | XP_006722901.1 | 12 | 3070 | |||
XM_011528173.1 | XP_011526475.1 | 12 | 2899 | |||
XM_011528174.2 | XP_011526476.1 | 12 | 3068 | |||
XM_017027111.1 | XP_016882600.1 | 11 | 3142 | |||
GenBank mRNA | AK294109.1 | - | 12, 13 | 1342 | ||
AL832550.1 | - | 11 | 2804 | |||
BC028101.1 | - | 12, 13 | 1545 | 5242909 |