Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044305.2 | NP_001037770.1 | 6 | 801 | ||
NM_001281439.1 | NP_001268368.1 | 5 | 720 | |||
NM_001281440.1 | NP_001268369.1 | 6 | 617 | |||
NM_021940.4 | NP_068759.2 | 5 | 720 | |||
XM_005248760.4 | XP_005248817.1 | 6 | 778 | |||
XM_011536045.2 | XP_011534347.1 | 6 | 778 | |||
XM_011536046.2 | XP_011534348.1 | 7 | 703 | |||
XM_011536047.2 | XP_011534349.1 | 5 | 697 | |||
XM_011536048.2 | XP_011534350.1 | 7 | 690 | |||
XM_017011170.1 | XP_016866659.1 | 7 | 886 | |||
GenBank mRNA | AF442495.1 | - | 5 | 618 | ||
AK023221.1 | - | 5 | 535 | |||
AK124239.1 | - | 7 | 734 | |||
AK222711.1 | - | 5 | 517 | |||
AK290448.1 | - | 6 | 578 | |||
AK307807.1 | - | 6 | 614 | |||
AK315579.1 | - | 5 | 598 | |||
AY055003.1 | - | 6 | 616 | |||
AY055004.1 | - | 5 | 535 | |||
BC028074.1 | - | 5 | 592 | 5215869 | ||
BC036123.1 | - | 6 | 654 | 5313112 |