Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021943.2 | NP_068762.1 | 6 | 1756 | ||
XM_005249269.1 | XP_005249326.1 | 5 | 1725 | |||
XM_011514790.1 | XP_011513092.1 | 5 | 1623 | |||
XM_017011171.1 | XP_016866660.1 | 5 | 1729 | |||
GenBank mRNA | AK023284.1 | - | 6 | 1431 | ||
AL137302.1 | - | 1 | 583 |