Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C14ORF93 SIRNA Assays ›
Gene Symbol:
C14orf93
Gene Name:
chromosome 14 open reading frame 93
Gene Aliases:
-
Chromosome Location:
Chr.14: 22986901 - 23010201 on Build GRCh38
UniGene ID:
Hs.255874
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133463
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C14orf93
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130706.2
NP_001124178.1
8
2173
NM_001130708.2
NP_001124180.1
7
2094
NM_001282968.1
NP_001269897.1
8
2028
NM_001282969.1
NP_001269898.1
8
1760
NM_001282970.1
NP_001269899.1
8
2220
NM_021944.3
NP_068763.2
7
2340
XM_005267971.1
XP_005268028.1
7
2242
XM_005267972.1
XP_005268029.1
7
2143
XM_006720231.1
XP_006720294.1
8
1808
XM_006720232.2
XP_006720295.1
9
1803
XM_011537059.1
XP_011535361.1
7
2326
XM_011537060.2
XP_011535362.1
8
2138
XM_011537062.1
XP_011535364.1
7
3395
XM_017021569.1
XP_016877058.1
8
6205
XM_017021570.1
XP_016877059.1
8
5490
XM_017021571.1
XP_016877060.1
9
5611
XM_017021572.1
XP_016877061.1
8
5450
XM_017021573.1
XP_016877062.1
8
2347
XM_017021574.1
XP_016877063.1
9
5405
XM_017021575.1
XP_016877064.1
9
5893
XM_017021576.1
XP_016877065.1
9
2039
XM_017021577.1
XP_016877066.1
9
4932
GenBank mRNA
AK022216.1
-
7
2282
AK023026.1
-
7
2027
BC014299.2
-
8
2139
3936989
CN309429.1
-
2
711
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-tm9kw:80/10.239.162.147:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0