Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPAS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193314.1 | NP_001180243.1 | 8 | 1091 | ||
NM_001193315.1 | NP_001180244.1 | 8 | 691 | |||
NM_001193316.1 | NP_001180245.1 | 7 | 544 | |||
NM_001193317.1 | NP_001180246.1 | 8 | 868 | |||
NM_022067.3 | NP_071350.2 | 9 | 962 | |||
XM_017021580.1 | XP_016877069.1 | 8 | 682 | |||
XM_017021581.1 | XP_016877070.1 | 8 | 816 | |||
GenBank mRNA | AK022769.1 | - | 8 | 639 | ||
AK022925.1 | - | 8 | 617 | |||
AK098254.1 | - | 8 | 1052 | |||
AK222706.1 | - | 8 | 637 | |||
AK297388.1 | - | 8 | 793 | |||
AK298354.1 | - | 7 | 490 | |||
BC015054.1 | - | 9 | 883 | 3920459 | ||
CR457294.1 | - | 7 | 545 |