Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040424.2 | NP_001035514.1 | 15 | 2063 | ||
NM_001282934.1 | NP_001269863.1 | 16 | 2123 | |||
NM_022115.4 | NP_071398.3 | 22 | 3050 | |||
NR_104257.1 | 16 | 2071 | ||||
NR_104258.1 | 15 | 1951 | ||||
NR_104260.1 | 15 | 1918 | ||||
NR_135464.1 | 15 | 2059 | ||||
XM_006724039.2 | XP_006724102.1 | 15 | 1905 | |||
XM_006724040.2 | XP_006724103.1 | 14 | 1845 | |||
XM_011529674.1 | XP_011527976.1 | 15 | 1935 | |||
XM_011529675.1 | XP_011527977.1 | 16 | 1979 | |||
XM_011529676.2 | XP_011527978.1 | 16 | 4388 | |||
XM_011529677.2 | XP_011527979.1 | 16 | 2064 | |||
XM_011529678.2 | XP_011527980.1 | 16 | 2108 | |||
XM_011529679.2 | XP_011527981.1 | 16 | 2441 | |||
XM_011529681.2 | XP_011527983.1 | 16 | 3345 | |||
XM_011529683.1 | XP_011527985.1 | 21 | 2766 | |||
XM_017028425.1 | XP_016883914.1 | 15 | 1918 | |||
XM_017028426.1 | XP_016883915.1 | 15 | 4327 | |||
GenBank mRNA | AB051812.1 | - | 15 | 2054 | ||
AB051813.1 | - | 16 | 2114 | |||
AB051814.1 | - | 16 | 2062 | |||
AB051815.1 | - | 15 | 1942 | |||
AY078498.2 | - | 22 | 3050 | |||
BC067102.1 | - | 15 | 1918 | 5925140 | ||
CF135664.1 | - | 2 | 146 | |||
CF137278.1 | - | 4 | 437 | |||
CN400247.1 | - | 3 | 321 |